Order Entry
Masterflex® Transfer Tubing, Kynar Flex™ 2750 PVDF, 1/4" ID x 3/8" OD; 50 ft
  MFLX95100-13
undefined
Masterflex® Transfer Tubing, Kynar Flex™ 2750 PVDF, 1/4" ID x 3/8" OD; 50 ft
  MFLX95100-13
 : MFLA95100-13
7 430,00 Kč
  1 * 50 Feet
  • Model:
  • 95100-13
  • 95100-09
  • 95100-11
  • 95100-05
  • 95100-07
  • 95100-15
  • 95100-19
  • 95100-01
  • 95100-03
  • 95100-21
  • Bend radius:
  • 25,4 mm (1")
  • 63,5 mm (2,5")
  • 12,7 mm (0,5")
  • 25,4 mm (1")
  • 44,45 mm (1,75")
  • 50,8 mm (2")
  • 127 mm (5")
  • 12,7 mm (0,5")
  • 19,05 mm (0,75")
  • 558,8 mm (22")
  • Clarity:
  • Translucent
  • Translucent
  • Translucent
  • Translucent
  • Translucent
  • Translucent
  • Translucent
  • Translucent
  • Translucent
  • Translucent
  • GMP compliant:
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Length per pack:
  • 15,24 m (50 ft.)
  • 15,24 m (50 ft.)
  • 15,24 m (50 ft.)
  • 15,24 m (50 ft.)
  • 15,24 m (50 ft.)
  • 15,24 m (50 ft.)
  • 15,24 m (50 ft.)
  • 15,24 m (50 ft.)
  • 15,24 m (50 ft.)
  • 15,24 m (50 ft.)
  • Max. pressure:
  • 12 bar (174 psi)
  • 5,79 bar (84 psi)
  • 17,93 bar (260 psi)
  • 8,69 bar (126 psi)
  • 6,96 bar (101 psi)
  • 9 bar (130 psi)
  • 6 bar (87 psi)
  • 31,1 bar (451 psi)
  • 11,58 bar (168 psi)
  • 4,48 bar (65 psi)
  • Max. temperature:
  • 135 °C (275 °F)
  • 135 °C (275 °F)
  • 135 °C (275 °F)
  • 135 °C (275 °F)
  • 135 °C (275 °F)
  • 135 °C (275 °F)
  • 135 °C (275 °F)
  • 135 °C (275 °F)
  • 135 °C (275 °F)
  • 135 °C (275 °F)
  • Min. temperature:
  • –40 °C (–40 °F)
  • –40 °C (–40 °F)
  • –40 °C (–40 °F)
  • –40 °C (–40 °F)
  • –40 °C (–40 °F)
  • –40 °C (–40 °F)
  • –40 °C (–40 °F)
  • –40 °C (–40 °F)
  • –40 °C (–40 °F)
  • –40 °C (–40 °F)
  • Regulatory approvals:
  • FDA;USP
  • FDA;USP
  • FDA;USP
  • FDA;USP
  • FDA;USP
  • FDA;USP
  • FDA;USP
  • FDA;USP
  • FDA;USP
  • FDA;USP
  • Sterilisation method EtO:
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Sterilisation method gamma:
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Working pressure:
  • 12 bar (174 psi)
  • 5,79 bar (84 psi)
  • 17,93 bar (260 psi)
  • 8,69 bar (126 psi)
  • 6,96 bar (101 psi)
  • 9 bar (130 psi)
  • 6 bar (87 psi)
  • 31,1 bar (451 psi)
  • 11,58 bar (168 psi)
  • 4,48 bar (65 psi)
  • Warranty:
  • Manufacturer defect
  • Manufacturer defect
  • Manufacturer defect
  • Manufacturer defect
  • Manufacturer defect
  • Manufacturer defect
  • Manufacturer defect
  • Manufacturer defect
  • Manufacturer defect
  • Manufacturer defect
  • Currently Viewing
Specifications
About this item
Excellent chemical resistance and UV stable.
  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory approvals to match your unique application
Applications: High purity and temperature, UV sensitive, or aggressive chemical fluid transfer.
Characteristics: Excellent chemical resistance, low TOC, rigid tubing works well with compression and over-molded fittings and adapters. Translucent.
Certification: FDA complaint (21 CFR 177.2600), meets USP Class VI requirements, and is made in accordance to cGMP.
Temperature range: -40 to 275 °F (-40 to 135 °C)
Sterilize: Gamma irradiation, or ethylene oxide.
  • Tubing ext.Ø:
    9,525 mm (0,375")
  • Tubing int.Ø:
    6,35 mm (0,25")
  • Wall thickness:
    1,588 mm
  • Max. pressure:
    12 bar (174 psi)
  • Max. temperature:
    135 °C (275 °F)
  • Colour:
    Translucent
  • Description:
    Transfer Tubing, Kynar Flex™ 2750 PVDF, 1/4" ID x 3/8" OD; 50 ft
  • Length:
    15,24 m
  • Seq:
    02
  • Environmentally Preferable:
  • Model:
    95100-13
  • Bend radius:
    25,4 mm (1")
  • GMP compliant:
    Yes
  • Length per pack:
    15,24 m (50 ft.)
  • Regulatory approvals:
    FDA;USP
  • Sterilisation method EtO:
    Yes
  • Sterilisation method gamma:
    Yes
  • Min. temperature:
    –40 °C (–40 °F)
  • Warranty:
    Manufacturer defect
  • Working pressure:
    12 bar (174 psi)
  • Clarity:
    Translucent
  • Tubing material:
    Kynar Flex™ 2750 PVDF
  • Pk:
    50 Feet