Capillary columns, VF-Xms
Dodavatel: VARIAN
Certifikáty
O tomto produktu
VF-Xms delivers the ultimate in sensitivity and signal-to-noise ratio, and is the low bleed, more polar alternative.
- High arylene modified phase for accurate results
- Isothermal applications up to 340 °C for a broad application range
- Ideal for confirmational analysis for ultimate confidence
VF-Xms comes with an EZ-GRIP, simplifying installation, coupling and operation of capillary columns. For guaranteed performance, the retention index, efficiency, selectivity and bleed is measured and specified on the test report supplied with each column.
Informace o produktu
- Catalog #Supplier #Column FormatPhaseFilm thicknessØ int.LengthPkDostupnostCena
- VARICP9041CP9041CapillaryVF-Xms0,15 µm0,15 mm20 m1Přihlásit se pro dostupnost1 * 1 KS23 260,00 KčSupplier Article Number: CP9041Local Article Number: VARICP9041Specifikace:
Specifications
Environmentally Preferable - Supplier Article Number: CP8801Local Article Number: VARICP8801Specifikace:
Specifications
Environmentally Preferable - VARICP8805CP8805CapillaryVF-Xms0,10 µm0,25 mm30 m1Přihlásit se pro dostupnost1 * 1 KS27 260,00 KčSupplier Article Number: CP8805Local Article Number: VARICP8805Specifikace:
Specifications
Environmentally Preferable - VARICP8806CP8806CapillaryVF-Xms0,25 µm0,25 mm30 m1Přihlásit se pro dostupnost1 * 1 KS27 260,00 KčSupplier Article Number: CP8806Local Article Number: VARICP8806Specifikace:
Specifications
Environmentally Preferable - VARICP8807CP8807CapillaryVF-Xms0,50 µm0,25 mm30 m1Přihlásit se pro dostupnost1 * 1 KS27 260,00 KčSupplier Article Number: CP8807Local Article Number: VARICP8807Specifikace:
Specifications
Environmentally Preferable - VARICP8813CP8813CapillaryVF-Xms0,25 µm0,32 mm30 m1Přihlásit se pro dostupnost1 * 1 KS29 410,00 KčSupplier Article Number: CP8813Local Article Number: VARICP8813Specifikace:
Specifications
Environmentally Preferable - VARICP8809CP8809CapillaryVF-Xms0,25 µm0,25 mm60 m1Přihlásit se pro dostupnost1 * 1 KS46 700,00 KčSupplier Article Number: CP8809Local Article Number: VARICP8809Specifikace:
Specifications
Environmentally Preferable - VARICP8816CP8816CapillaryVF-Xms0,25 µm0,32 mm60 m1Přihlásit se pro dostupnost1 * 1 KS50 800,00 KčSupplier Article Number: CP8816Local Article Number: VARICP8816Specifikace: