This knife cut cleanroom wipe made of 100% continuous filament polyester knit fabric provides high levels of cleanliness, abrasion resistance and chemical compatibility.
- Low particles, fibers, ions and extractables
- High abrasion resistance
- Chemically compatible with IPA, Acetone and other solvents
- Light weight, high sorbency
- Low endotoxin levels
- Soft hand and edge for sensitive surfaces
Ideal for integrated circuit fabrication areas, CD and DVD manufacturing operations and stencil and other print roll cleaning applications. Also suitable for equipment wrap and wipe downs.
Suitable for use in Class 10, ISO Class 4, M2.5 cleanrooms.
Colour: White
Upozornení: Data accurate at time of publishing, latest test results and test methods can be found on supplier's website.