Chcete se opět přihlásit?
Absorbent mats, oil-only, 4 in 1®, PIG®
Dodavatel: NEW PIG
PIG® PIG® Oil-Only 4 in 1®
NEWPMAT435EA 4350 CZK InStock
NEWPMAT435 NEWPMAT484
Absorbent mats, oil-only, 4 in 1®, PIG®
Absorpční materiály
Highly absorbent mats are made from a eight-layer 100% PP medium-weight construction and function as pads, rolls, socks or wipes. They absorb and retain a variety of leaks, drips, and spills of oils and fuels, except water.
  • Scuff-resistant, tear-resistant, even under heavy foot traffic or abrasive conditions
  • Flame-resistant - material melts when exposed to high heat
  • Dimple pattern wicks of liquid throughout mat for faster and easier clean-up
  • Rolls are perforated every 250 mm
Objednat
OBJEDNÁVKA
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Nebo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Popis Katalogové číslo Availability Jednotka Vaše cena Price Per Množství