HPLC columns, NUCLEOSIL® C18 AB
NUCLEOSIL® C18 AB is a specially crosslinked octadecyl modified silica phase for HPLC.
- Robust spherical silica gel
- Strongly hydrophobic, endcapped phase with high steric selectivity for RP chromatography
- Separation mechanism based on hydrophobic and steric interactions
EC guard columns are used for analytical EC columns with the column protection system guard column holder (554-5851).
- Catalog No:
- MANA720936.40
- MANA720305.46
- MANA720936.46
- MANA721073.30
- MANA720935.40
- MANA720935.46
- Carbon load:
- 25%
- 25%
- 25%
- 25%
- 25%
- 25%
- Endcapped:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- pH range:
- 1 - 9
- 1 - 9
- 1 - 9
- 1 - 9
- 1 - 9
- 1 - 9
- USP code:
- L1
- L1
- L1
- L1
- L1
- L1
Analytical columns
- Column FormatPhasePore sizeParticle SizeØ int.Length
- MANA720936.40720936.40AnalyticalC18 AB100 Å5 µm4,00 mm250 mm1 * 1 KS19 790,00 Kč
Specifications
Environmentally PreferableCarbon load25%EndcappedYespH range1 - 9USP codeL1 - MANA720305.46720305.46AnalyticalC18 AB100 Å5 µm4,60 mm150 mm1 * 1 KS18 180,00 Kč
Specifications
Environmentally PreferableCarbon load25%EndcappedYespH range1 - 9USP codeL1 - MANA720936.46720936.46AnalyticalC18 AB100 Å5 µm4,60 mm250 mm1 * 1 KS21 080,00 Kč
Specifications
Environmentally PreferableCarbon load25%EndcappedYespH range1 - 9USP codeL1 - MANA720935.40720935.40AnalyticalC18 AB100 Å5 µm4,00 mm125 mm1 * 1 KS17 380,00 Kč
Specifications
Environmentally PreferableCarbon load25%EndcappedYespH range1 - 9USP codeL1 - MANA720935.46720935.46AnalyticalC18 AB100 Å5 µm4,60 mm125 mm1 * 1 KS18 180,00 Kč
Specifications
Environmentally PreferableCarbon load25%EndcappedYespH range1 - 9USP codeL1
Guard columns
- Column FormatPhasePore sizeParticle SizeØ int.Length
- MANA721073.30721073.30Guard columnC18 AB100 Å5 µm3,00 mm4 mm1 * 3 KS4 930,00 Kč
Specifications
Environmentally PreferableCarbon load25%EndcappedYespH range1 - 9USP codeL1