Tisk…
Gauntlets, dry box
Dodavatel: Piercan
PIEC18800CA106/10EA 10190 CZK
PIEC18800CA106/10 PIEC18800CA86/10 PIEC13800CA86/10 PIEC18800CA96/10 PIEC18750CA96/10
Gauntlets, dry box
Neoprene, anti-static
EN 388 (X111), EN 374 (ACK), EN 421; Cat. III