Disposable Particle Filtering Respirators, FFP1/FFP2/FFP3, Classic
Supplier: Moldex
Certificates
About this item
Product Information
- Catalog #Supplier #ClassWith valveTypePkAvailabilityPrice
- 111-1142236015FFP1-2360201 * 20 KS1 160,00 KčSupplier Article Number: 236015Local Article Number: MOLD236015Specifications:
Specifications
Environmentally Preferable - 111-0525236515FFP1+2365201 * 20 KS2 120,00 KčSupplier Article Number: 236515Local Article Number: MOLD236515Specifications:
Specifications
Environmentally Preferable - 111-05322400FFP2-2400201 * 20 KS2 130,00 KčSupplier Article Number: 2400Local Article Number: MOLD2400Specifications:
Specifications
Environmentally Preferable - 111-05332405FFP2+2405201 * 20 KS3 060,00 KčSupplier Article Number: 2405Local Article Number: MOLD2405Specifications:
Specifications
Environmentally Preferable - 111-11432555FFP3+2555201 * 20 KS5 030,00 KčSupplier Article Number: 2555Local Article Number: MOLD2555Specifications: