Tisk…
Round tissue float bath
Dodavatel: GFL
631-1300EA 13640 CZK
GFLA1052
Round tissue float bath
The tissue float bath has a black anodised aluminium interior, the housing is powder coated aluminium. The unit has precise temperature regulation which ensures evenly stretched paraffin wax sections, and allows the sections to be easily seen in the bath or on the rim of the bath when drying.
  • Low height (100 mm) enables comfortable and safe working
  • Knob and scale temperature regulator
  • Control thermometer in stainless steel holder is fixed to the rim of the bath
Informace pro objednávání: Supplied without aluminium lid which protects against dust.