Chcete se opět přihlásit?
Bis(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')calcium, Sigma-Aldrich®
Dodavatel: Sigma-Aldrich
SAFA362956-1GEA 0 CZK InStock
SAFA362956-1G
Bis(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')calcium, Sigma-Aldrich®
Bis(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')calcium
Vzorec: C₂₂H₃₈CaO₄
MDL Number: MFCD00064757
CAS číslo: 36818-89-0
Objednat
OBJEDNÁVKA
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Nebo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Popis Katalogové číslo Availability Jednotka Vaše cena Price Per Množství