Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')terbium(III), Sigma-Aldrich®
Dodavatel: Merck
SIAL434051-1GEA
3330
CZK
SIAL434051-1G
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')terbium(III), Sigma-Aldrich®
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')terbium(III)
Vzorec:
C₃₃H₅₇O₆Tb MW: 708,727 g/mol |
MDL Number:
MFCD00064752 CAS číslo: 15492-51-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...