Copper(II) hexafluoroacetylacetonate hydrate, Sigma-Aldrich®
Dodavatel: Merck
SIAL335193-5GEA
5170
CZK
SIAL335193-5G
Copper(II) hexafluoroacetylacetonate hydrate, Sigma-Aldrich®
Copper(II) hexafluoroacetylacetonate hydrate
Vzorec:
C₁₀H₂CuF₁₂O₄·xH₂O MW: 477,64 g/mol (anhy) |
MDL Number:
MFCD00151019 CAS číslo: 155640-85-0 EINECS: 238-850-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...