Ethyl-3-phenylglycidate (mixture of cis and trans isomers), Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
SIAL291404-100MLEA
1650
CZK
SIAL291404-100ML
Ethyl-3-phenylglycidate (mixture of cis and trans isomers), Sigma-Aldrich®
Ethyl-3-phenylglycidate (mixture of cis and trans isomers)
Vzorec:
C₁₁H₁₂O₃ MW: 192,21 g/mol |
MDL Number:
MFCD00005123 CAS číslo: 121-39-1 EINECS: 204-467-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...