Tisk…
Lithium diphenylphosphide 0.5 mol/l in THF, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
SIAL464872-100MLEA 0 CZK
SIAL464872-100ML
Lithium diphenylphosphide 0.5 mol/l in THF, Sigma-Aldrich®
Lithium diphenylphosphide
Vzorec: C₁₂H₁₀LiP
MDL Number: MFCD00046054
CAS číslo: 65567-06-8