Tisk…

5-Chloro-2,1,3-benzothiadiazol-4-ylamine

TCIAA1513-5GEA 8200 CZK
TCIAA1513-5G
5-Chloro-2,1,3-benzothiadiazol-4-ylamine
5-Chloro-2,1,3-benzothiadiazol-4-ylamine
Vzorec: C₆H₄ClN₃S
MDL Number: MFCD00465908
CAS číslo: 30536-19-7
EINECS: 457-490-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR