Tisk…

1-(2-Tetrahydrofuroyl)piperazine hydrobromide, 97%

H55490.06EA 2640 CZK
ALFAH55490.06 ALFAH55490.14 ALFAH55490.03
1-(2-Tetrahydrofuroyl)piperazine hydrobromide, 97%
Purity: 97%
CAS No.: 63590-62-5
Molecular Formula: C9H16N2O2.HBr
Formula Weight: 265.15
Physical Form: Powder
Appearance: Off-white
Melting Point: 103-107°
MDL No.: MFCD03701619
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR