Isosorbide dimethyl ether ≥98.0% (by GC)
TCIAI0390-25GEA
2260
CZK
TCIAI0390-25G
Isosorbide dimethyl ether ≥98.0% (by GC)
Isosorbide dimethyl ether
Vzorec:
C₈H₁₄O₄ MW: 174,20 g/mol Bod vzplanutí: 108°C |
MDL Number:
MFCD00064828 CAS číslo: 5306-85-4 EINECS: 226-159-8 |
Výsledky specifikačního testu
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Specific rotation [a]20/D | +96.0 to +99.0 deg (C=2,chloroform) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...