Tisk…

2-Amino-5-methyl-4-phenylthiazole, Sigma-Aldrich®

Dodavatel: Sigma-Aldrich

SAFA658480-10GEA 0 CZK
SAFA658480-10G SAFA658480-1G
2-Amino-5-methyl-4-phenylthiazole, Sigma-Aldrich®
2-Amino-5-methyl-4-phenylthiazole
Vzorec: C₁₀H₁₀N₂S
MDL Number: MFCD00617257
CAS číslo: 30709-67-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR