Tisk…

Ethyl 4-methyl-2-(4-pyridyl)thiazole-5-carboxylate, 97%

H54601.03EA 1440 CZK
ALFAH54601.03 ALFAH54601.MD ALFAH54601.06
Ethyl 4-methyl-2-(4-pyridyl)thiazole-5-carboxylate, 97%
Purity: 97%
CAS No.: 89401-54-7
Molecular Formula: C12H12N2O2S
Formula Weight: 248.30
Melting Point: 58-62°
MDL No.: MFCD06739813
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR