Tisk…
4-Methyl-2-(4-pyridyl)thiazole-5-carboxylic acid, 97+%
H51753.06EA 7310 CZK
ALFAH51753.06 ALFAH51753.03
4-Methyl-2-(4-pyridyl)thiazole-5-carboxylic acid, 97+%
Purity: 97+%
CAS No.: 144060-98-0
Molecular Formula: C10H8N2O2S
Formula Weight: 220.25
Melting Point: 302-307°
MDL No.: MFCD00171757