Tisk…

6-Bromo-1H-[1,2,3]triazolo[4,5-b]pyridine

Dodavatel: TCI EUROPE

TCIAB3034-1GEA 4180 CZK
TCIAB3034-1G TCIAB3034-5G
6-Bromo-1H-[1,2,3]triazolo[4,5-b]pyridine
6-Bromo-1H-[1,2,3]triazolo[4,5-b]pyridine
Vzorec: C₅H₃BrN₄
MDL Number: MFCD04060716
CAS číslo: 92276-38-5
https://cz.vwr.com/stibo/web/std.lang.all/44/76/28934476.jpg

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR