Tisk…
N-(3-Fluorobenzyl)-3,4-dimethylaniline, 97%
H57956.MDEA 814 CZK
ALFAH57956.MD ALFAH57956.03
N-(3-Fluorobenzyl)-3,4-dimethylaniline, 97%
Purity: 97%
CAS No.: 1019612-14-6
Molecular Formula: C15H16FN
Formula Weight: 229.30
MDL No.: MFCD11144364