Tisk…

1,4-Diamino-2,3-dicyano-9,10-anthraquinone

Dodavatel: TCI EUROPE

TCIAD2148-25GEA 0 CZK
TCIAD2148-25G
1,4-Diamino-2,3-dicyano-9,10-anthraquinone
1,4-Diamino-2,3-dicyano-9,10-anthraquinone
Vzorec: C₁₆H₈N₄O₂
MDL Number: MFCD00053082
CAS číslo: 81-41-4
EINECS: 201-347-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR