Tisk…

Bis(trimethylsilyl)methane

TCIAB1857-10MLEA 6810 CZK
TCIAB1857-10ML
Bis(trimethylsilyl)methane
Bis(trimethylsilyl)methane
Vzorec: C₇H₂₀Si₂
MDL Number: MFCD00008275
CAS číslo: 2117-28-4
EINECS: 218-322-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR