Tisk…

Tris(trimethylsilyl)methane

Dodavatel: ABCR GMBH

ABCRAB109332.0025EA 0 CZK
ABCRAB109332.0025
Tris(trimethylsilyl)methane
Tris(trimethylsilyl)methane
Vzorec: C₁₀H₂₈Si₃
MW: 232,59 g/mol
MDL Number: MFCD00008273
CAS číslo: 1068-69-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR