Tisk…

6,8-Dibromo-3-formylchromone, Sigma-Aldrich®

Dodavatel: Sigma-Aldrich

SAFA402176-1GEA 0 CZK
SAFA402176-1G
6,8-Dibromo-3-formylchromone, Sigma-Aldrich®
6,8-Dibromo-3-formylchromone
Vzorec: C₁₀H₄Br₂O₃
MDL Number: MFCD00191991
CAS číslo: 42059-76-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR