Tisk…

6-Ethyl-3-formylchromone, Sigma-Aldrich®

Dodavatel: Sigma-Aldrich

SAFA402133-1GEA 0 CZK
SAFA402133-1G
6-Ethyl-3-formylchromone, Sigma-Aldrich®
6-Ethyl-3-formylchromone
Vzorec: C₁₂H₁₀O₃
MDL Number: MFCD00192155
CAS číslo: 42059-78-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR