Tisk…
Methyldiphenylphosphine oxide, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ200
SIAL454427-5GEA 0 CZK
SIAL454427-5G SIAL454427-25G
Methyldiphenylphosphine oxide, Sigma-Aldrich®
Methyldiphenylphosphine oxide
Vzorec: C₁₃H₁₃OP
MW: 216,22
MDL Number: MFCD00002081
CAS číslo: 2129-89-7
EINECS: 218-346-8