Tisk…

1-Bromo-3-(trimethylsilyl)benzene, Sigma-Aldrich®

Dodavatel: Sigma-Aldrich

SAFA714798-10GEA 0 CZK
SAFA714798-10G
1-Bromo-3-(trimethylsilyl)benzene, Sigma-Aldrich®
1-Bromo-3-(trimethylsilyl)benzene
Vzorec: C₉H₁₃BrSi
MDL Number: MFCD00093882
CAS číslo: 17878-47-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR