Tisk…

3-Bromo-1-propylboronic acid pinacol ester, Sigma-Aldrich®


M-Clarity™ quality level = MQ100

SIAL695858-1GEA 0 CZK
SIAL695858-1G
3-Bromo-1-propylboronic acid pinacol ester, Sigma-Aldrich®
3-Bromo-1-propylboronic acid pinacol ester
Vzorec: C₉H₁₈BBrO₂
MW: 248,96 g/mol
MDL Number: MFCD10567053
CAS číslo: 124215-44-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR