Tisk…

Potassium 2-phenylethyltrifluoroborate, Sigma-Aldrich®

Dodavatel: Sigma-Aldrich

SAFA563099-1GEA 0 CZK
SAFA563099-1G
Potassium 2-phenylethyltrifluoroborate, Sigma-Aldrich®
Potassium 2-phenylethyltrifluoroborate
Vzorec: C₈H₉BF₃K
MW: 212,06 g/mol
MDL Number: MFCD09039257
CAS číslo: 329976-74-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR