Tisk…

4-(Trimethylsilyl)phenylboronic acid, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

SIAL523674-5GEA 0 CZK
SIAL523674-5G SIAL523674-1G
4-(Trimethylsilyl)phenylboronic acid, Sigma-Aldrich®
4-(Trimethylsilyl)phenylboronic acid
Vzorec: C₉H₁₅BO₂Si
MDL Number: MFCD00093348
CAS číslo: 17865-11-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR