Tisk…

4-Bromo-N-cyclopentylbenzamide, 97%

H57511.MDEA 1530 CZK
ALFAH57511.MD ALFAH57511.03
4-Bromo-N-cyclopentylbenzamide, 97%
Purity: 97%
CAS No.: 223557-21-9
Molecular Formula: C12H14BrNO
Formula Weight: 268.15
MDL No.: MFCD00588998
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR