Tisk…

3,4-Diaminobenzhydrazide, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

SIAL579750-5GEA 4410 CZK
SIAL579750-5G
3,4-Diaminobenzhydrazide, Sigma-Aldrich®
3,4-Diaminobenzhydrazide
Vzorec: C₇H₁₀N₄O
MW: 166,18 g/mol
MDL Number: MFCD00017069
CAS číslo: 103956-09-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR