Tisk…

N-Cyclohexyl-N-methyl-2-fluorobenzamide, 97%

H56688.MDEA 1230 CZK
ALFAH56688.MD ALFAH56688.03
N-Cyclohexyl-N-methyl-2-fluorobenzamide, 97%
Purity: 97%
CAS No.: 1841-37-8
Molecular Formula: C14H18FNO
Formula Weight: 235.30
MDL No.: MFCD01214100
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR