Tisk…

Potassium 2-(hydroxymethyl)phenyltrifluoroborate, 96%

H53193.06EA 1830 CZK
ALFAH53193.06
Potassium 2-(hydroxymethyl)phenyltrifluoroborate, 96%
Purity: 96%
CAS No.: 850623-74-4
Molecular Formula: C7H7BF3KO
Formula Weight: 214.04
Melting Point: 300°
MDL No.: MFCD04115769
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR