Tisk…
2-Isopropoxy-5-(trifluoromethyl)benzeneboronic acid, 96%
H53121.03EA 3990 CZK
ALFAH53121.03 ALFAH53121.MD
2-Isopropoxy-5-(trifluoromethyl)benzeneboronic acid, 96%
Purity: 96%
CAS No.: 850593-12-3
Molecular Formula: C10H12BF3O3
Formula Weight: 248.01
Melting Point: 66-68°
MDL No.: MFCD07363786