Tisk…
2-Bromo-N-(2-fluorobenzyl)-4-methylaniline, 97%
H57697.MDEA 2040 CZK
ALFAH57697.MD ALFAH57697.03
2-Bromo-N-(2-fluorobenzyl)-4-methylaniline, 97%
Purity: 97%
CAS No.: 1019565-10-6
Molecular Formula: C14H13BrFN
Formula Weight: 294.16