Tisk…
3-Chloro-N-(2-fluorobenzyl)-2-methylaniline, 97%
Dodavatel: Thermo Scientific
H57529.03EA 3060 CZK
ALFAH57529.03 ALFAH57529.MD
3-Chloro-N-(2-fluorobenzyl)-2-methylaniline, 97%
Purity: 97%
CAS No.: 1040321-18-3
Molecular Formula: C14H13ClFN
Formula Weight: 249.71