Tisk…
5-Chloro-N-(2-fluorobenzyl)-2-methylaniline, 97%
H57964.MDEA 2040 CZK
ALFAH57964.MD ALFAH57964.03
5-Chloro-N-(2-fluorobenzyl)-2-methylaniline, 97%
Purity: 97%
CAS No.: 1036483-41-6
Molecular Formula: C14H13ClFN
Formula Weight: 249.71