Tisk…

4-Chloro-N-(4-fluorobenzyl)-3-methylaniline, 97%

H57655.03EA 2330 CZK
ALFAH57655.03 ALFAH57655.MD
4-Chloro-N-(4-fluorobenzyl)-3-methylaniline, 97%
Purity: 97%
Molecular Formula: C14H13ClFN
Formula Weight: 249.71
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR