Tisk…

4-tert-Butyl-N-(4-fluorobenzyl)aniline, 97%

H57714.MDEA 814 CZK
ALFAH57714.MD ALFAH57714.03
4-tert-Butyl-N-(4-fluorobenzyl)aniline, 97%
Purity: 97%
CAS No.: 1041549-32-9
Molecular Formula: C17H20FN
Formula Weight: 257.35
MDL No.: MFCD11923957
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR