Tisk…

2-Fluoro-N-(4-fluorobenzyl)-3-methylaniline, 97%

H57958.MDEA 1750 CZK
ALFAH57958.MD ALFAH57958.03
2-Fluoro-N-(4-fluorobenzyl)-3-methylaniline, 97%
Purity: 97%
Molecular Formula: C14H13F2N
Formula Weight: 233.26
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR