Tisk…

3-Fluoro-N-(2-fluorobenzyl)-5-methylaniline, 97%

H57570.03EA 8140 CZK
ALFAH57570.03 ALFAH57570.MD
3-Fluoro-N-(2-fluorobenzyl)-5-methylaniline, 97%
Purity: 97%
Molecular Formula: C14H13F2N
Formula Weight: 233.26
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR