Tisk…

3-Fluoro-N-(3-fluorobenzyl)-4-methylaniline, 97%

H57713.03EA 4440 CZK
ALFAH57713.03 ALFAH57713.MD
3-Fluoro-N-(3-fluorobenzyl)-4-methylaniline, 97%
Purity: 97%
CAS No.: 1019493-00-5
Molecular Formula: C14H13F2N
Formula Weight: 233.26
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR