Tisk…

3-Fluoro-N-(4-fluorobenzyl)-2-methylaniline, 97%

H57362.MDEA 1020 CZK
ALFAH57362.MD ALFAH57362.03
3-Fluoro-N-(4-fluorobenzyl)-2-methylaniline, 97%
Purity: 97%
CAS No.: 1282172-35-3
Molecular Formula: C14H13F2N
Formula Weight: 233.26
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR