Tisk…
4-Fluoro-N-(3-fluorobenzyl)-2-methylaniline, 97%
H57039.MDEA 2450 CZK
ALFAH57039.MD ALFAH57039.03
4-Fluoro-N-(3-fluorobenzyl)-2-methylaniline, 97%
Purity: 97%
CAS No.: 1021079-97-9
Molecular Formula: C14H13F2N
Formula Weight: 233.26