Tisk…

3-Hydroxybenzaldehyde azine, Supelco®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

17031-1GEA 0 CZK
17031-1G
3-Hydroxybenzaldehyde azine, Supelco®
3-Hydroxybenzaldehyde azine
Vzorec: C₁₄H₁₂N₂O₂
MDL Number: MFCD00557128
CAS číslo: 18428-76-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR