Tisk…

Potassium 2-benzyloxyphenyltrifluoroborate, 95%

H52686.03EA 3480 CZK
ALFAH52686.03 ALFAH52686.MD
Potassium 2-benzyloxyphenyltrifluoroborate, 95%
Purity: 95%
CAS No.: 850623-44-8
Molecular Formula: C13H11BF3KO
Formula Weight: 290.13
Melting Point: 200-204°
MDL No.: MFCD04115742
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR