Tisk…

2,4-Dibromo-N-(2-fluorobenzyl)aniline, 97%

H57481.MDEA 642 CZK
ALFAH57481.MD ALFAH57481.03
2,4-Dibromo-N-(2-fluorobenzyl)aniline, 97%
Purity: 97%
CAS No.: 1152619-64-1
Molecular Formula: C13H10Br2FN
Formula Weight: 359.03
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR