Tisk…

4-Bromo-3-fluoro-N-(2-fluorobenzyl)aniline, 97%

H57720.MDEA 872 CZK
ALFAH57720.MD ALFAH57720.03
4-Bromo-3-fluoro-N-(2-fluorobenzyl)aniline, 97%
Purity: 97%
Molecular Formula: C13H10BrF2N
Formula Weight: 298.13
MDL No.: MFCD21009057
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR